Flint Glass Gas Measuring Tubes
Fournisseur: ROCHESTER APPAREL & AD SPECIALTIES
Certificats
A propos de cet article
These fine flint glass tubes measure the volume of gas generated and absorbed by substances undergoing chemical action.
The top is closed and the tube is graduated from the top downward.
Customers Who Bought This Also Bought