Flint Glass Clear Glass Plates
Fournisseur: F J Gray & Co.
Certificats
A propos de cet article
These double thick, glass plates have seamed edges and are useful in covering gas bottles, beakers, and other glassware
Packaging: Packages of 12.
Customers Who Bought This Also Bought