Flex-Tanks and Covers
Fournisseur: Nasco
Certificats
A propos de cet article
Versatile Flex-Tanks and covers may be used as aquariums, terrariums, or critter chambers.
- Vented covers
- Versatile
- Not autoclavable
- Made of thermo plastic
Tanks do not crack or break.
Customers Who Bought This Also Bought