Flexible Magnetic Strips
# de catalogue 470006-802
Fournisseur: Skywill Far East Industries
Spécifications
- Description:Flexible Magnetic Strips
- Cat. no.:470006-802
- Supplier No.:6611800
Spécifications
A propos de cet article
Use these flexible magnetic strips for experiments, to hold objects in place, or for creating your own inventions.
They are easily trimmed to any size with scissors. The strips are 25×1.6 mm thick. 3 m roll.
Customers Who Bought This Also Bought