Flat Flexible SAFE-T® Ruler
# de catalogue 470046-496
Fournisseur: Learning Resources
Spécifications
- Color:Clear
- Description:Flat and flexible Safe-T ruler
- Cat. no.:470046-496
- Length:300 mm (12")
- Supplier No.:43060
Spécifications
A propos de cet article
Projects a clear, sharp image on the overhead. May be used for all other ruler activities.
U.S. Standard inch and metric calibrations. Durable View-Thru™ SAFE-T® Plastic
Customers Who Bought This Also Bought