Fizz Wizard and Jammin’ Jelly Reaction
# de catalogue 470145-532
Fournisseur: Universe of Science
Spécifications
- Description:Fizz Wizard and Jammin’ Jelly Reaction
- Cat. no.:470145-532
- Supplier No.:2100-B
Spécifications
A propos de cet article
Chemistry experiments you can drink and eat. Make soda pop and make it change colors.
Pull tasty jelly things from a solution. Make Polymer Pudding. Learn about chemical reactions, acidbase reactions, indicators, pH and gels. Note: This kit contains food grade materials.
Kit contains: Magic Color Crystals (cabbage extract), sodium bicarbonate, citric acid, calcium acetate, sodium alginate, pipet, spoon, small scoops, cups, stirrers and instructions.
Caution: Do not attempt these experiments with any other chemical samples.
Customers Who Bought This Also Bought