Spécifications
- Description:Firearm Identification Kit
- No. of Lab Groups:6
- Cat. no.:470133-328
- Supplier No.:BF-01
Spécifications
A propos de cet article
Practice Real-World Firearm Investigation
Students will be challenged and engaged as they incorporate math and logic to investigate and identify bullets and firearms. Using the same procedures as forensic crime laboratories, students will determine types of bullets and the possible firearms that could have fired them. A detailed PowerPoint is included to enhance the realistic samples used in demonstration.
- Identification of Bullets
- Use of Math and Logic in Forensic activities
Students will be challenged and engaged as they incorporate math and logic to investigate and identify bullets and firearms. Using the same procedures as forensic crime laboratories, students will determine types of bullets and the possible firearms that could have fired them. A detailed PowerPoint is included to enhance the realistic samples used in demonstration.