Fingerprint Lifting Strips
# de catalogue 470190-892
Fournisseur: PHILPAC CORPORATION
Spécifications
- Description:Fingerprint Lifting Strips
- Cat. no.:470190-892
- Supplier No.:DITA-00033
Spécifications
A propos de cet article
These individually cut and backed strips make lifting latent prints of various sizes extremely easy.
- Each pad comes with 25 sheets
- Size of each: 4 x 6"
These individually cut and backed strips make lifting latent prints of various sizes extremely easy. No cutting is necessary, so there’s no need to worry about finding the loose end on your roll of tape. Each pad comes with 25 sheets. Size of each: 4" x 6".
Customers Who Bought This Also Bought