Fingerprint Ink Strips
# de catalogue 470014-882
Fournisseur: MELLEN MARKETING ASSOCIATES
Spécifications
- Description:Fingerprint Ink Strips
- Cat. no.:470014-882
- Supplier No.:229
Spécifications
A propos de cet article
Perfect Prints Every Time
- Size: 1" x 5"
- Package of 200
Simply peel back the covering on these pre-inked strips and you’re ready to start taking fingerprints. They have just the right amount of ink to get great prints with minimum mess, and the ink washes off easily. Size: 1" x 5". Package of 200.
Customers Who Bought This Also Bought