Filter Support Collar
# de catalogue 470017-722
Fournisseur: United Scientific Supplies
Spécifications
- Description:Filter Support Collar
- Cat. no.:470017-722
- Supplier No.:FLTR01
Spécifications
A propos de cet article
Filter Support Collar is a sturdy, molded rubber filter disc.
- Inside Diameter: 32 mm
- Outside Diameter: 76 mm
- Molded Rubber Construction
It is designed to support and seal a filter crucible or funnel in the neck of a filter flask.
Customers Who Bought This Also Bought