Fiber Optic Element
# de catalogue 470006-588
Fournisseur: Arcor Electronics
Spécifications
- Description:Fiber Optic Element, 3 Ft Length, 1/16" Width
- Cat. no.:470006-588
- Supplier No.:FOK-1
Spécifications
A propos de cet article
3' Fiber optic element, easily demonstrates the bending of light through the fiber optic wire. An inexpensive way to introduce students to fiber optics specifically the use of fiber optics for visual applications.
Teacher notes explain what fiber optic is and how it works and offers suggestions for activities. An economical way to prepare students for more advanced fiber optic studies/kits.
Customers Who Bought This Also Bought