Fever Scan Tape
# de catalogue 470012-694
Fournisseur: Liquid Crystal Resources
Spécifications
- Description:Thermometer, LCD Temperature Tape (Fever Scan)
- Cat. no.:470012-694
- Supplier No.:FT-55U
Spécifications
A propos de cet article
Pressed against the forehead for 35 to 40 seconds, this fever tester indicates body temperature.
- Range: 95 – 104 °F
Customers Who Bought This Also Bought