BoneClones® Fetal Skull Set
Fournisseur: BONE CLONES, INC.
Certificats
A propos de cet article
Compare Human Fetal Skulls At Various Stages Of Development
- Show skull development from 20 weeks to full term development
- High quality casts of real specimens
An illustrative aid for any anatomy, child development, or parenting course, these replica skulls show multiple stages of development. Demonstrate the growth of fetal bones and compare the fontanelles at each stage. Also available as a set of 5 skulls at 14, 20, 24, 30, and 34-weeks. To prevent damage the jaws are not removable from these models.
Ordering information: Each skull is shipped with an osteological evaluation report.
Customers Who Bought This Also Bought