GPI Anatomicals® Feline Heart and Lung Model
Fournisseur: GPI ANATOMICALS
Certificats
A propos de cet article
Show The Effect Of Heartworm On Household Pets
Average feline heart and lungs infested with a single heartworm (Dirofilaria immitis). Cut-away view demonstrates structures in the heart (right ventricle, pulmonary trunk, pulmonary arteries) and lungs where adult-stage heartworms are normally found. The cut section of the lung indicates patches of heartworm-induced irritation.
Customers Who Bought This Also Bought