Faraday Cage
# de catalogue 470039-428
Fournisseur: Science First
Spécifications
- Description:Faraday Cage
- Cat. no.:470039-428
- Supplier No.:10-094
Spécifications
A propos de cet article
Demonstrate that a charge cannot exist inside a conductor and re-create Faraday’s classic 'Ice pail' experiment with this kit.
- With instructions
- Ø×H: 12×27 cm
Delivery: Includes a perforated metal cage with stand and cover as well as a mounted metal point for the lightning rod effect.
Customers Who Bought This Also Bought