Extension Condenser Clamps with Swivel Jaws
Catalog # 470148-704
Supplier: VWR International
Specifications
- Description:Clamp, Condenser, 50 mm Jaw Opening, w/ Vinyl Coated Swivel Jaws, Extension, Humboldt
- Cat. no.:470148-704
- Supplier No.:CS2921/1
Specifications
About this item
This clamp features vinyl-coated swivel jaws made from stamped steel.
The jaws can be set at any angle to the forged steel rod, which measures 9.5 mm in diameter. 50 mm jaw opening.
Customers Who Bought This Also Bought