Euplectella Aspergillum
# de catalogue 470225-114
Fournisseur: Avantor
Spécifications
- Length:25.4 - 30.4 cm (10 - 12")
- Description:Glass Sponge, Dry Preserved
- Cat. no.:470225-114
- Supplier No.:WARD470225-114
Spécifications
A propos de cet article
Modern glass sponge known as "Venus Flower Basket".
- Porifera
- Dehydrated Preserved
- 10 to 12" long
- Phillipines
Customers Who Bought This Also Bought