Essential Physics Demo: Large Tuning Forks
# de catalogue 470124-122
Fournisseur: VWR International
Spécifications
- Description:Essential Physics Demo: Large Tuning Forks
- Cat. no.:470124-122
- Supplier No.:DEMO-TFK
Spécifications
A propos de cet article
Lower Frequency Than Traditional Forks
The large tines on these tuning forks create visible vibrations and emit sound waves at a lower frequency than traditional tuning forks. Set of two forks includes a larger fork with 50cm tines at 12Hz, and a smaller fork with 37cm tines at 38Hz. Also includes a detailed demonstration guide.
- Includes a detailed demonstration guide.
The large tines on these tuning forks create visible vibrations and emit sound waves at a lower frequency than traditional tuning forks. Set of two forks includes a larger fork with 50cm tines at 12Hz, and a smaller fork with 37cm tines at 38Hz. Set of two.
Customers Who Bought This Also Bought