ContactUsLinkComponent
Essential Physics Demo: Rotating Speaker Bar
Essential Physics Demo: Rotating Speaker Bar
# de catalogue 470124-124
Fournisseur:  VWR International
SPEAKER BAR ROTATING PHYSICS DEMO
Essential Physics Demo: Rotating Speaker Bar
# de catalogue 470124-124
Fournisseur:  VWR International
Spécifications
  • Description:
    Essential Physics Demo: Rotating Speaker Bar
  • Cat. no.:
    470124-124
  • Supplier No.:
    RMB-2010
Spécifications
A propos de cet article
Experiment With Sound Wave Interference
Affordable, Easy, Fun
Present these demonstrations in front of your class to excite your students about learning core physics concepts. Set up and execution are simple, so you can spend more time engaging your students. Ideal for introducing a unit or using as reinforcement after a lesson, these demos are available for all fundamental physical science topics.
Demonstrate the Doppler effect and the concept of sound interference with this oscillating speaker bar. Create active participation among the entire class as students mobilize around the classroom and listen as changes in movable objects affect sound quality. When the speaker bar and students are stationary, the sound will be constant. When the bar rotates and students remain put or choose to roam the room, varying interference patterns will be heard. As interference patterns change from constructive to destructive, overall sound quality will change. Includes a detailed demonstration guide. Requires a connecting cord and function generator, not included.
  • Includes a detailed demonstration guide.
Affordable, Easy, Fun
Present these demonstrations in front of your class to excite your students about learning core physics concepts. Set up and execution are simple, so you can spend more time engaging your students. Ideal for introducing a unit or using as reinforcement after a lesson, these demos are available for all fundamental physical science topics.
Demonstrate the Doppler effect and the concept of sound interference with this oscillating speaker bar. Create active participation among the entire class as students mobilize around the classroom and listen as changes in movable objects affect sound quality. When the speaker bar and students are stationary, the sound will be constant. When the bar rotates and students remain put or choose to roam the room, varying interference patterns will be heard. As interference patterns change from constructive to destructive, overall sound quality will change. Requires a connecting cord and function generator, not included.
Customers Who Bought This Also Bought