Equilibrium Tubes
# de catalogue 470005-472
Fournisseur: VWR International
Spécifications
- Description:Equilibrium Tubes w/ Support
- Cat. no.:470005-472
- Supplier No.:PH0200B
Spécifications
A propos de cet article
A set of four borosilicate glass tubes of different shapes connected together to illustrate that liquids will stand at the same level.
The four glass tubes are sealed to a manifold and mounted on a support.
Customers Who Bought This Also Bought