ContactUsLinkComponent
Equal Mass Set, Eisco
Equal Mass Set, Eisco
# de catalogue 470017-032
Fournisseur:  VWR International
EQUAL MASS SET 15 G SET/5
Equal Mass Set, Eisco
# de catalogue 470017-032
Fournisseur:  VWR International
Spécifications
  • Description:
    Equal Mass Set
  • Cat. no.:
    470017-032
  • Supplier No.:
    PH0115-5
Spécifications
A propos de cet article
Five-piece equal mass density set – for specific heat experiments and more.
  • Set includes aluminum, brass, nylon, acrylic and polyvinyl chloride
  • The five cylinders in this set have an equal mass, measure 12.5 mm (0.5”) in diameter, but vary in length from 0.5 to 4”
Excellent for physics classrooms. Demonstrate the inverse relationship between mass and density with this equal mass set. Density materials have a wide range of educational applications for all grade levels, include density investigations, specific heat and specific gravity exploration. Set includes a complete instruction manual that gives a background on density, specific gravity and specific heat and includes inquiry based student activities with capture sheets.
Customers Who Bought This Also Bought