Epoxy Cement
Catalog # 470020-090
Supplier: ELLSWORTH ADHESIVE
Specifications
- Description:Epoxy Cement
- Cat. no.:470020-090
- Supplier No.:154509
Specifications
About this item
This is an epoxy cement kit.
- Kit includes both resin and hardener tubes
- Resin: 2 ½ oz.
- Hardener: ¾ oz.
The epoxy will cure in a day, or in just hours at higher temperatures.
Customers Who Bought This Also Bought