ContactUsLinkComponent
Environmental Test Kits
Environmental Test Kits
# de catalogue 470105-474
Fournisseur:  Science First
CLEAN WATER TEST KIT
Environmental Test Kits
# de catalogue 470105-474
Fournisseur:  Science First
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Test Kit, How Clean is the Soil Kit
  • Cat. no.:
    470105-474
  • Supplier No.:
    5800
Spécifications
A propos de cet article
Help students gain a better awareness of environmental conditions in their area and in the world at large.
  • Examine the components which make up soil
  • Demonstrate the impact of soil pollution
  • Includes instruction guide
The physical makeup of soil, including nitrate, phosphate, potassium, chlorides, oil and microorganisms, is explored in How Clean Is the Soil? Thermal and Sewage Pollution demonstrates how microorganisms and chemicals contribute to water pollution. All Kits include an instruction guide.
Customers Who Bought This Also Bought