ContactUsLinkComponent
Envirolab 1
Envirolab 1
# de catalogue 470004-428
Fournisseur:  Theta Technologies
ENVIROLAB I LAB ACTIVITY
Envirolab 1
# de catalogue 470004-428
Fournisseur:  Theta Technologies
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    EnviroLab 1
  • No. of Lab Groups:
    12
  • Cat. no.:
    470004-428
  • Supplier No.:
    360950
Spécifications
A propos de cet article
Geologic Chemistry Applications Solve Hypothetical Scenario
  • Test for heavy metals and organic compounds
  • Linked to a real world scenario
  • Includes a teacher's guide
Carterville is a small town with a big problem. The town has grown astronomically over the last hundred years, but never as fast as it has within the last ten. Now the pains of growth have struck the town, its citizens, businesses and most importantly, Carter Creek. In this ecological simulation, students join the Environmental Protection Services to solve the mystery of the pollution of Carter Creek. The students perform chemical tests on both the water and sediment from the creek. Once the tests have been performed they decide who or what is responsible for the fouling of the creek. All tests are simulations of real tests for lead, mercury, PCBs and 2,4 D (2, 4-Dichlorophenoxyacetic Acid). Includes a teacher’s guide.
Customers Who Bought This Also Bought