Energy Transformation Kit
# de catalogue 470148-756
Fournisseur: VWR International
Spécifications
- Description:Kit, Energy Transformation
- Cat. no.:470148-756
- Supplier No.:P20360A
Spécifications
A propos de cet article
Appropriate for grades 6 to 12 Brings the First Law of Thermodynamics to life! This pair of precision-machined, chrome-plated steel balls is actually a hands-on demonstration in energy transformation. By striking the balls together, you will create enough heat at the point of contact to burn a hole in a sheet of paper.
There will be no flame but a charred hole will appear instantly along with the smell of burning paper. Each ball weighs 454 g. Includes instructions with suggestions for tying the activity into national science standards.
Customers Who Bought This Also Bought