ContactUsLinkComponent
Ward's® Endospore Staining Kit
Ward's® Endospore Staining Kit
# de catalogue 470030-202
Fournisseur:  Avantor
WARDS ENDOSPORE STAINING KIT
Ward's® Endospore Staining Kit
# de catalogue 470030-202
Fournisseur:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Endospore Staining Kit
  • Cat. no.:
    470030-202
Spécifications
A propos de cet article
Examine a non-pathogenic bacterial strain that illustrates endospore formation.
  • Demonstration plate culture of Bacillus cereus
Under environmentally stressful conditions, vegetative bacteria enter a heat-resistant stage that helps them survive; this stage is known as an endospore.
The bacterial culture and stain in this kit provide a striking example of this stage.
The kit includes:
A coupon redeemable for a plate culture of Bacillus cereus.
Blank microscope slides and applicator swabs
Staining solutions
Canada: Containment Level 2 – PHAC License required.
Ordering information: This item contains a coupon for live or perishable materials. Please provide a date that the live or perishable components will arrive.
Delivery: The kit will ship separately from the live component. Kits do not contain perishable components and generally ship via UPS ground. Perishable components will ship via overnight or second day delivery.
Customers Who Bought This Also Bought