Prepared Eosin Methylene Blue Agar (EMB) Media
# de catalogue 470194-720
Fournisseur: IPM SCIENTIFIC, INC.
Numéro CAS:
Spécifications
- Packaging:Plates
- Description:EMB Media
- Cat. no.:470194-720
- Supplier No.:880960
Spécifications
A propos de cet article
This selective and differential medium is used to isolate gram negative organisms.
- Sterile, pre-prepared media; just inoculate and incubate.
- Selective and differential for gram negative organisms
- Contains indicator dyes that form purple precipitate at low pH
This media contains Eosin Y and Methylene Blue, which are indicator dyes, forming a dark purple precipitate at a low pH, an indication of the presence of gram negative organisms such as fecal coliforms. This medium is not meant to act as a general purpose medium and will not support the growth of gram positive organisms.
Each bottle pours approximately 10 Petri dishes. Simply melt in a microwave oven, water bath or autoclave. Once cool enough to handle, pour into sterile Petri dish plates
Customers Who Bought This Also Bought