Elodea, Submerged Leaf Slide
# de catalogue 470177-460
Fournisseur: Avantor
Spécifications
- Description:Elodea, Submerged Leaf
- Cat. no.:470177-460
- Stain:Quadruple stained
- Supplier No.:WARD470177-460
- Orientation:Cross section
Spécifications
A propos de cet article
Illustrates Aquatic Plant Adaptations
- Identifying features clearly distinguishable
- Rigorous quality control standards
Waterweed. Hydrophytic leaf.
Customers Who Bought This Also Bought