Electromagnet Science Set
# de catalogue 470013-330
Fournisseur: DOWLING MAGNETS,LLC
Spécifications
- Description:Electromagnet Science Set
- Cat. no.:470013-330
- Supplier No.:SDK300
Spécifications
A propos de cet article
Observe Magnets At Work In Everyday Life
- Create science projects with everyday materials
- All materials included
Build a doorbell, telegraph system, or even a catapult by using a true electromagnet. This set allows students to create working science projects using simple materials such as batteries, nails, and light bulbs. It comes with all necessary materials and instructions.
Caution: WARNING: This product contains a small magnet(s). Swallowed magnets can stick together across intestines causing serious infections and death. Seek immediate medical attention if magnet(s) are swallowed or inhaled.
Customers Who Bought This Also Bought