Spécifications
- Description:Electrode, Electrolysis, w/o Leads
- Cat. no.:470105-768
- Supplier No.:6678000
Spécifications
A propos de cet article
The two electrodes are constructed of stainless steel and have curved ends flattened at the tips.
A special plastic covering coats the rod up to the binding area. The electrodes are available with or without connecting leads. We recommend the leads and clips to simplify connections.