Electric Plume
# de catalogue 470003-972
Fournisseur: Science First
Spécifications
- Description:Electric Plume, Accessory for Electrostatic Generator (Van de Graaff)
- Cat. no.:470003-972
- Supplier No.:N-125
Spécifications
A propos de cet article
Graphically illustrate the fact that like charges repel each other.
The multi-colored ribbons are connected to a suction cup that sits on top of a Van de Graaff generator. As the generator operates, the ribbons accumulate a charge and stand on end.
Customers Who Bought This Also Bought