Electric Heating Mantles
Fournisseur: Glas-Col
Certificats
A propos de cet article
Designed to Operate Below Red Heat at Maximum Input Voltage.
- Made entirely of glass fabric
- Hemispherical construction allows full view of flask contents while heating
Mantles provide evenly distributed heat which may be closely controlled by adjustment of the input voltage. (Heating Mantle Controller sold separately WLS-40883 / 470202-750)
With separable 2-wire line cord, 4' L. For 115VAC or DC. CSA listed.
Certifications: CSA listed.
Customers Who Bought This Also Bought