Electric Generator
# de catalogue 470006-534
Fournisseur: Jensen Manufacturing
Spécifications
- Description:Generator, Electric
- Cat. no.:470006-534
- Supplier No.:JENSEN STEAM ENGINE MFG.CO.INC
Spécifications
A propos de cet article
This belt-driven generator may be used with our Steam Engine (63030-00 ). Operating at approximately 1500 rpm, the generator will produce enough electricity to light the included 3.8 volt bulb. The unit is mounted on a metal base (7.6×11.4 cm) and has a V-grooved pulley for the drive belt.
Customers Who Bought This Also Bought