Optical Lens Cleaner, Aldon
# de catalogue 470153-310
Fournisseur: Ward's Science
Spécifications
- Description:Lens cleaner, spray, optical
- Cat. no.:470153-310
- Supplier No.:LL0158-8OZ
Spécifications
A propos de cet article
Cleans without leaving residue.
- Store between 40 to 100 °F
Our concentrated formula cleans glass or hard plastic safely, quickly and effectively. Simply apply and wipe clean. Unlike similar fluids, it cleans without leaving a residue, does not exhibit an annoying odor, and does not dry too quickly, making it ideal for precision lens, window, or prism cleaning.
Customers Who Bought This Also Bought