ContactUsLinkComponent
Economy Wave Generator
Economy Wave Generator
# de catalogue 470110-486
Fournisseur:  Science First
ECONOMY WAVE GENERATOR
Economy Wave Generator
# de catalogue 470110-486
Fournisseur:  Science First
Spécifications
  • Description:
    Economy Wave Generator
  • Cat. no.:
    470110-486
  • Supplier No.:
    15445
Spécifications
A propos de cet article
Ruggedly constructed
  • Includes two point sources for generating circular waves, phase control for varying interference patterns, ripple bar for generating straight waves, and variable amplitude and frequency
Includes instructions. Requires a variable low voltage power supply: 1.5-3V, not included.
Customers Who Bought This Also Bought