Economy Wave Generator
# de catalogue 470110-486
Fournisseur: Science First
Spécifications
- Description:Economy Wave Generator
- Cat. no.:470110-486
- Supplier No.:15445
Spécifications
A propos de cet article
Ruggedly constructed
- Includes two point sources for generating circular waves, phase control for varying interference patterns, ripple bar for generating straight waves, and variable amplitude and frequency
Includes instructions. Requires a variable low voltage power supply: 1.5-3V, not included.
Customers Who Bought This Also Bought