Economy Short Stem Funnels, Hutzler
Fournisseur: Hutzler Manufacturing
Certificats
A propos de cet article
Funnels are made of durable plastic. Funnels of top inner diameter 70-108 are polypropylene and autoclavable. Funnels with top inner diameter of 127 and 165 are polyethylene and not autoclavable.
The 165 mm size has a wide stem for quick transfers.
Customers Who Bought This Also Bought