Economy Rod-Mounted Pulley
Catalog # 470149-504
Supplier: VWR International
Specifications
- Description:Economy Rod-Mounted Pulley
- Cat. no.:470149-504
- Supplier No.:P10317A
Specifications
About this item
Excellent for Introductory Mechanics Studies
- Low friction cone bearings ensure accurate, repeatable results
Consists of a 10cm-long rod securely fastened to a durable nylon fork. The nylon sheave is 4.5cm in diameter.
Customers Who Bought This Also Bought