Economy Lab Ovens
Fournisseur: Quincy Labs
Certificats
A propos de cet article
Heavy Steel Double-Wall Construction.
- Includes dual range laboratory thermometer and three wire cord/plug
- 115 VAC, 50/60 Hz
- Constructed with UL and CSA listed components
These gravity convection ovens are for drying, baking, sterilizing, evaporating and testing. Maximum temperature is 220°C. Ovens feature corrosive-resistant aluminized steel interior, perforated heat shield for good temperature uniformity and sheathed heating elements for quick run-up and recovery times. Features light gray, hard, scratch-resistant hammer finish with two adjustable chrome plated wire shelves and supports.
Customers Who Bought This Also Bought