ContactUsLinkComponent
Bucket and Cylinder
Bucket and Cylinder
# de catalogue 470110-368
Fournisseur:  VWR International
ECONOMY ARCHIMEDES PRINCIPLE
Bucket and Cylinder
# de catalogue 470110-368
Fournisseur:  VWR International
Spécifications
  • Description:
    Bucket and Cylinder
  • Cat. no.:
    470110-368
  • Supplier No.:
    PH0120A
Spécifications
A propos de cet article
Made of brass, bucket with suspension loop and hook.
  • For demonstrating Archimedes's principle
Simple form in brass, suitable for class demonstration. The bucket has suspension loop at top and hook at the bottom. The cylinder has a suspension hook at one end. Size of 6 cm long, overall length when extended 16 cm.
Customers Who Bought This Also Bought