Easy Science Demos And Labs: Life Science Book
Catalog # 470022-670
Supplier: Walch Publishing
Specifications
- Description:Easy Science Demos And Labs: Life Science Book
- Length:21 cm
- Cat. no.:470022-670
- No. of Pages:116
- Width:27 cm
- Supplier No.:0-45015
Specifications
About this item
Thomas Kardos
Make your life science lessons memorable with visual demonstrations sure to capture students’ attention. Using only common items already in your classroom and lab, you can show students a variety of plant and animal processes. The book also includes materials lists, safety tips, a glossary, and an index.
Customers Who Bought This Also Bought