Dynamo
# de catalogue 470120-948
Fournisseur: VWR International
Spécifications
- Description:Dynamo
- Cat. no.:470120-948
- Supplier No.:PH1245-N8
Spécifications
A propos de cet article
The Dynamo illustrates how electricity is produced.
- Hand-Wheel Apparatus Generates Current
- Two Lamp Holders and Bulbs Represent Currents
- Wooden Base Attached to Apparatus
- Process Shows How Electricity Is Produced
The hand-wheel apparatus generates alternating and direct current.
Two lamp holders and bulbs represent currents, and the apparatus is attached to a polished wooden base.
Customers Who Bought This Also Bought