Order Entry
ContactUsLinkComponent
Duck Skull, Ward’s®
Duck Skull, Ward’s®
# de catalogue 470027-994
Fournisseur:  SKULLS UNLIMITED
SKULL DUCK NATURAL BONE
Duck Skull, Ward’s®
# de catalogue 470027-994
Fournisseur:  SKULLS UNLIMITED

Spécifications

  • Description:
    Duck Skull
  • Height:
    1.5"
  • Cat. no.:
    470027-994
  • Length:
    5"
  • Width:
    11/8"
  • Supplier No.:
    SB-010 A
  • No. of Parts:
    1

Spécifications

A propos de cet article

Compare this skull of an adult domestic duck, family Anatidae, to other domestic fowl.

  • Natural bone
  • Professionally cleaned and prepared
Ordering information: This is a real specimen, so size and appearance may vary from the image.
Customers Who Bought This Also Bought