Boreal Science Drosophila Recessive Genes Set
# de catalogue 470189-394
Fournisseur: Boreal Laboratories
Spécifications
- Description:Live Drosophila Culture, Recessive Genes Set
- Cat. no.:470189-394
- Supplier No.:WARD470189-394
Spécifications
A propos de cet article
The set Includes cultures of wild, white, brown, dumpy, vestigial, vestigial brown and sepia.
We fully guarantee the purity and viability of our Drosophila melanogaster cultures. A large, vigorous culture is shipped in a shatterproof plastic vial. To ship Drosophila cultures with living larvae we require 10 days' notice prior to use date. During very cold weather, adult flies may die but eggs and larvae usually survive. Allow cultures to come to room temperature before checking under low magnification for eggs and live larvae. Complete instructions are always included.
Customers Who Bought This Also Bought