Dropping Bottle Block
# de catalogue 470123-936
Fournisseur: Bennett Wood Specialties
Spécifications
- Description:Dropping Bottle Block
- Cat. no.:470123-936
- Supplier No.:6793 + PKG
Spécifications
A propos de cet article
Holds six 30 mL round dropping bottles. Oil treated wood block with rubber feet, drilled with holes 1-1/2" dia. Size: 12" W x 2-1/4" D. Without bottles.
Customers Who Bought This Also Bought