Disposable Polyethylene Dropper Pipettes
# de catalogue 470152-908
Fournisseur: Avantor
Spécifications
- Length:150 mm
- Description:Beral pipette, graduated, disposable polyethylene, 6572900
- Cat. no.:470152-908
- Stem Diameter:5 mm
- Supplier No.:WARD470152-908
Spécifications
A propos de cet article
Chemically inert polyethylene pipettes.
Designed for consistent bulb pressure and tip circumference to ensure precisely measured drop reproducibility.
Customers Who Bought This Also Bought