Double-Walled Calorimeter
# de catalogue 470018-276
Fournisseur: PRECISION MOULDED POLYMERS LIMITED
Spécifications
- Description:Double-Walled Calorimeter
- Cat. no.:470018-276
- Supplier No.:M40101
Spécifications
A propos de cet article
Simple Calorimeter to Measure Heat in Many Applications
The double-walled cover minimizes heat loss, and the inner and outer vessels are both made of aluminum. The inner vessel, which has a 200 mL capacity, comes with a stirrer and support.
Customers Who Bought This Also Bought