Double-sided Dry Erase Mats
# de catalogue 470046-830
Fournisseur: Learning Resources
Spécifications
- Description:Double-sided Dry Erase Mats
- Cat. no.:470046-830
- Supplier No.:39143
Spécifications
A propos de cet article
These economical double-sided plastic coated mats feature a 1 cm grid with x-y axis bolded for easy graphing activities.
- Size of each: 9×11"
Use dry or wet erase markers to plot along each axis; the grid squares allow students to easily visualize the relationships between numbers and space on the graph. The other side of the mat is blank for your own activities. and both sides can be used with dry or wet erase markers.
Packaging: Pack of 10.
Customers Who Bought This Also Bought