Dogfish Dissection Tray
# de catalogue 470004-500
Fournisseur: Nasco
Small Business Enterprise
Spécifications
- Description:Dogfish Dissection Tray, Tray
- Cat. no.:470004-500
- Supplier No.:SB14284DIM
Spécifications
A propos de cet article
This lightweight, chemically resistant polyethylene tray holds even the largest dogfish specimen, ventral side up, because the molded central channel and grove hold the dorsal fin and spine in place.
It also features a nonskid bottom. Size: 31"L×151/2"W×15/8"H.
Ordering information: The clear plastic cover is available separately.
Customers Who Bought This Also Bought