DNA SuperModel Kit
# de catalogue 470029-868
Fournisseur: RYLER ENTERPRISES, INC.
Spécifications
- Description:DNA Supermodel Kit
- No. of Lab Groups:1
- Cat. no.:470029-868
- Supplier No.:DNA-1
Spécifications
A propos de cet article
Re-Create Accurate Bonding Between Base Pairs
Easy to set up, this model is flexible, durable, and economical. The colorful components provide an accurate representation of bonding between base pairs by showing the double bonds between adenine and thymine and the triple bonds between guanine and cytosine. The kit comes with 183 pieces, which is enough to make a 24 nucleotide sequence. Includes an information and instruction booklet.
Caution: WARNING: CHOKING HAZARD - Small parts. Not for children under 3 yrs.
Customers Who Bought This Also Bought