ContactUsLinkComponent
DNA Fingerprinting by Restriction Enzyme Patterns
DNA Fingerprinting by Restriction Enzyme Patterns
# de catalogue 470200-476
Fournisseur:  Edvotek
KT LAB DNA FINGERPRINTING 1 EDVOTEK
DNA Fingerprinting by Restriction Enzyme Patterns
# de catalogue 470200-476
Fournisseur:  Edvotek
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    DNA Fingerprinting I Lab Activity
  • No. of Lab Groups:
    8
  • Cat. no.:
    470200-476
  • Supplier No.:
    109
Spécifications
A propos de cet article
In this experiment, crime scene DNA fingerprints are compared with those obtained from suspects.
  • Required Components Included
  • Time Required: 45 minutes
  • Kit Explains Basic Concepts of DNA Fingerprinting
DNA fingerprinting is a method used in various forensic procedures as well as paternity determinations. The experiment provides the basic reagents and biologicals for DNA fragmentation pattern analysis and does not contain human DNA.
Horizontal gel electrophoresis apparatus, DC power supply, and micropipettes with tips are required but not included.
Customers Who Bought This Also Bought